Kyselina kaprylová ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Octanoic acid, n-Octanoic Acid, Octoic acid
A11149.0F
A11149.AC
A11149.AP
A11149.0FEA
3090
CZK
A11149.0F
A11149.AC
A11149.AP
Kyselina kaprylová ≥98%
Kyselina kaprylová
Purity: 98+%
CAS No.: 124-07-2
Molecular Formula: C8H16O2
Formula Weight: 144.21
Flash Point: 132°(269°F)
Physical Form: Liquid
Density: 0.910
Refractive Index: 1.4275
Appearance: Colourless
Odour: Rancid
pH: 4
Melting Point: 16-17°
Boiling Point: 236-238°
Beilstein Reference: 1747180
Merck Reference: 14,1765
UN No.: 3265
MDL No.: MFCD00004429
Vzorec:
C₈H₁₆O₂ MW: 144,21 g/mol Bod varu: 236…238 °C Bod tání: 16…17 °C Hustota: 0,910 Bod vzplanutí: 132 °C (269 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004429 CAS číslo: 124-07-2 EINECS: 204-677-5 UN: 3265 ADR: 8,III Merck Index: 14,1765 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...