Tisk…

Kyselina pelargonová ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Nonanoic acid, Pelargonic Acid

B21568.AK B21568.AU
B21568.AKEA 961 CZK
B21568.AK B21568.AU
Kyselina pelargonová ≥97%
Kyselina pelargonová

Purity: 97% CAS No.: 112-05-0 Molecular Formula: CH3(CH2)7CO2H Formula Weight: 158.24 Flash Point: 140°(284°F) Refractive Index: 1.4315 Odour: Unpleasant pH: 3.5 Melting Point: 10-12° Boiling Point: 253-255° Beilstein Reference: 1752351 Merck Reference: 14,7070 UN No.: 3265 MDL No.: MFCD00004433

Vzorec: C₉H₁₈O₂
MW: 158,24 g/mol
Bod varu: 253…255 °C
Bod tání: 10…12 °C
Hustota: 0,905
Bod vzplanutí: 140 °C (284 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004433
CAS číslo: 112-05-0
EINECS: 203-931-2
UN: 3265
ADR: 8,III
Merck Index: 14,7070

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR