Heptanoic acid ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Heptylic acid, Enanthic acid
A17704.0F
A17704.AP
A17704.0FEA
4640
CZK
A17704.0F
A17704.AP
Heptanoic acid ≥98%
Heptanoic acid
Purity: 98+%
CAS No.: 111-14-8
Molecular Formula: C7H14O2
Formula Weight: 130.19
Flash Point: 114°(237°F)
Refractive Index: 1.4230
Melting Point: -8°
Boiling Point: 222-223°
Beilstein Reference: 1744723
Merck Reference: 14,4660
UN No.: 3265
MDL No.: MFCD00004426
Vzorec:
C₇H₁₄O₂ MW: 130,19 g/mol Bod varu: 222…223 °C Bod tání: –8 °C Hustota: 0,917 Bod vzplanutí: 114 °C (237 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004426 CAS číslo: 111-14-8 EINECS: 203-838-7 UN: 3265 ADR: 8,III Merck Index: 14,4660 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...