Tisk…

Kyselina octová ledová ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

33252.AK 33252.K2 33252.M4
33252.AKEA 1420 CZK
33252.AK 33252.K2 33252.M4
Kyselina octová ledová ≥99%
Kyselina octová

Purity: 99+% CAS No.: 64-19-7 Molecular Formula: CH3CO2H Formula Weight: 60.05 Flash Point: 40°(104°F) Physical Form: Liquid Density: 1.049 Refractive Index: 1.3721 Odour: Pungent pH: 2,5 Melting Point: 16.6° Boiling Point: 118.1° Merck Reference: 14,55 UN No.: 2789 MDL No.: MFCD00036152

Vzorec: H₃CCOOH
Bod varu: 118,1 °C
Hustota: 1,049
Bod vzplanutí: 40 °C (104 °F)
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS číslo: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
Merck Index: 14,55
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR