Tisk…

Arachidic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Eicosanylic acid, Arachidinic acid, Icosanoic acid, Eicosanoic acid

M-Clarity™ quality level = MQ200

A3631-1G A3631-10G A3631-25G A3631-500MG A3631-5G
SIALA3631-1GEA 1770 CZK
SIALA3631-1G SIALA3631-10G SIALA3631-25G SIALA3631-500MG SIALA3631-5G
Arachidic acid, Sigma-Aldrich®
Arachidic acid
Vzorec: C₂₀H₄₀O₂
MW: 312.54
Bod varu: 328
Bod tání: 75.5
Hustota: 0.824 g/cm³ (100 °C) (lit.)
Storage Temperature: Freezer
MDL Number: MFCD00002755
CAS číslo: 506-30-9
EINECS: 208-031-3
Merck Index: 12,00802

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR