Arachidic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Eicosanylic acid, Arachidinic acid, Icosanoic acid, Eicosanoic acid
10930-1G
10930-25G
10930-5G
SIAL10930-1GEA
1290
CZK
SIAL10930-1G
SIAL10930-25G
SIAL10930-5G
Arachidic acid, Sigma-Aldrich®
Arachidic acid
Vzorec:
C₂₀H₄₀O₂ MW: 312.54 Bod varu: 328 Bod tání: 75.5 Hustota: 0.824 g/cm³ (100 °C) (lit.) Storage Temperature: Freezer |
MDL Number:
MFCD00002755 CAS číslo: 506-30-9 EINECS: 208-031-3 Merck Index: 12,00802 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...