Tisk…

Kyselina laurová ≥99.5%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Dodecanoic acid, Dodecylic acid

42038.18 42038.30 42038.A1
42038.18EA 1130 CZK
42038.18 42038.30 42038.A1
Kyselina laurová ≥99.5%
Kyselina laurová

Purity: 99.5% CAS No.: 143-07-7 Molecular Formula: CH3(CH2)10CO2H Formula Weight: 200.32 Flash Point: 165°(329°F) Physical Form: Crystalline Density: 0.883 Odour: Odourless Melting Point: 43-45° Boiling Point: 180°/21mm Beilstein Reference: 1099477 Merck Reference: 14,5384 MDL No.: MFCD00002736

Vzorec: C₁₂H₂₄O₂
MW: 200.32
Bod varu: 180 °C (21 mmHg)
Bod tání: 43…45 °C
Hustota: 0.875
Bod vzplanutí: 165 °C (329 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002736
CAS číslo: 143-07-7
EINECS: 205-582-1
Merck Index: 13,05400

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR