Amylacetát ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
n-Pentyl acetate, Acetic acid amyl ester, Acetic acid pentyl ester, Pentyl acetate
L10852.22
L10852.36
L10852.22EA
865
CZK
L10852.22
L10852.36
Amylacetát ≥99%
Amylacetát
Purity: 99%
CAS No.: 628-63-7
Molecular Formula: CH3CO2(CH2)4CH3
Formula Weight: 130.19
Flash Point: 24°(75°F)
Refractive Index: 1.4030
Odour: Fruit-like
Melting Point: -100°
Boiling Point: 142°
Beilstein Reference: 1744753
UN No.: 1104
MDL No.: MFCD00009500
Vzorec:
C₇H₁₄O₂ Bod varu: 142 °C Bod tání: –100 °C Hustota: 0,876 Bod vzplanutí: 24 °C (75 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009500 CAS číslo: 628-63-7 EINECS: 211-047-3 UN: 1104 ADR: 3,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...