Octyl acetate ≥98%
Dodavatel: Thermo Scientific
Varování
|
Synonyms:
Acetic acid octyl ester
L06405.18
L06405.30
L06405.18EA
917
CZK
L06405.18
L06405.30
Octyl acetate ≥98%
Octyl acetate
Purity: 98+%
CAS No.: 112-14-1
Molecular Formula: C10H20O2
Formula Weight: 172.27
Flash Point: 86°(186°F)
Refractive Index: 1.4190
Boiling Point: 210-211°
Beilstein Reference: 1754554
MDL No.: MFCD00009562
Vzorec:
C₁₀H₂₀O₂ MW: 172,27 g/mol Bod varu: 210…211 °C Bod tání: –38,5 °C Hustota: 0,868 Bod vzplanutí: 86 °C (186 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009562 CAS číslo: 112-14-1 EINECS: 203-939-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...