Ethylacetát ≥99.5%, Spectrophotometric Grade for spectrophotometry
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Acetic acid ethyl ester
39177.K2
39177.M1
39177.K2EA
2720
CZK
39177.K2
39177.M1
Ethylacetát ≥99.5%, Spectrophotometric Grade for spectrophotometry
Ethylacetát
Purity: 99.5+%
CAS No.: 141-78-6
Molecular Formula: C4H8O2
Formula Weight: 88.11
Flash Point: -3°(26°F)
Physical Form: Liquid
Density: 0.902
Refractive Index: 1.3720
Appearance: Colourless
Odour: Sweet
Melting Point: -84°
Boiling Point: 77°
Beilstein Reference: 506104
Merck Reference: 14,3757
UN No.: 1173
MDL No.: MFCD00009171
Vzorec:
CH₃COOC₂H₅ MW: 88.11 Bod varu: 77 °C Bod tání: –84 °C Hustota: 0.902 Bod vzplanutí: –3 °C (26 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009171 CAS číslo: 141-78-6 EINECS: 205-500-4 UN: 1173 ADR: 3 II REACH: 01-2119475103-46 Merck Index: 14,3757 |
|
Výsledky specifikačního testu
Residual water | 0.10% |
Impurity content: Chlorinated pesticides | 5 ppt |
Residue on Evaporation | 3 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...