Tisk…

Decyl acetate ≥98%

Dodavatel: Thermo Fisher Scientific

A18814.0B A18814.30
A18814.0BEA 3190 CZK
A18814.0B A18814.30
Decyl acetate ≥98%
Decyl acetate

Purity: 98% CAS No.: 112-17-4 Molecular Formula: CH3CO2(CH2)9CH3 Formula Weight: 200.32 Flash Point: 125°(257°F) Refractive Index: 1.4270 Boiling Point: 244° Beilstein Reference: 1762123 MDL No.: MFCD00026538

Vzorec: C₁₂H₂₄O₂
Bod varu: 244 °C
Hustota: 0,867
Bod vzplanutí: 125 °C (257 °F)
Storage Temperature: Ambient
MDL Number: MFCD00026538
CAS číslo: 112-17-4
EINECS: 203-942-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR