Diacethyl 99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
2,3-Butanedione
107650050
107651000
107655000
ACRO107650050EA
1300
CZK
ACRO107650050
ACRO107651000
ACRO107655000
Diacethyl 99%
Diacethyl
Vzorec:
C₄H₆O₂ MW: 86.09 Bod varu: 190 Bod tání: 4…2 °C Hustota: 0.985 g/cm³ Bod vzplanutí: 7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008756 CAS číslo: 431-03-8 EINECS: 207-069-8 UN: 2346 ADR: 3 II Merck Index: 13,02985 |
Výsledky specifikačního testu
Appearance | Clear yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3910 to 1.3990 (20°C, 589 nm) |
GC | ≥98.5 % |
Specific gravity | (20°C) 0.98 to 0.99 |
Suitability | (for Cu + Fe determination) Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...