3-Mercaptobutanone ≥98% stabilizovaný
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
2-Mercapto-3-butanone, 3-Mercapto-2-butanone
Stabilization:
Stabilised with calcium carbonate
B22494.06
B22494.14
B22494.06EA
1570
CZK
B22494.06
B22494.14
3-Mercaptobutanone ≥98% stabilizovaný
3-Mercaptobutanone
Stabilized with 0.1% Calcium carbonate
Purity: 98%
CAS No.: 40789-98-8
Molecular Formula: C4H8OS
Formula Weight: 104.17
Storage Temperature: -30°C to -10°C
Flash Point: 47°(117°F)
Refractive Index: 1.4750
Boiling Point: 50°/18mm
UN No.: 1224
MDL No.: MFCD00085206
Vzorec:
C₄H₈OS MW: 104.17 Bod varu: 50 °C (18 mmHg) Hustota: 1.015 Bod vzplanutí: 47 °C (117 °F) Storage Temperature: Freezer |
MDL Number:
MFCD00085206 CAS číslo: 40789-98-8 EINECS: 255-082-2 UN: 1224 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...