Tisk…

Aceton ≥99.5%, Environmental Grade for environmental analysis

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

43053.K2 43053.K7 43053.M6
43053.K2EA 1640 CZK
43053.K2 43053.K7 43053.M6
Aceton ≥99.5%, Environmental Grade for environmental analysis
Aceton

Purity: 99.5+% CAS No.: 67-64-1 Molecular Formula: C3H6O Formula Weight: 58.08 Flash Point: -17°(1°F) Physical Form: Liquid Density: 0.791 Refractive Index: 1.3590 Appearance: Colourless Odour: Characteristic Melting Point: -94° Boiling Point: 56° Beilstein Reference: 635680 Merck Reference: 14,66 UN No.: 1090 MDL No.: MFCD00008765


Suitable for pesticide residue analysis

Vzorec: CH₃COCH₃
Bod varu: 56 °C
Bod tání: –94 °C
Hustota: 0,791
Bod vzplanutí: –17 °C (1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS číslo: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 14,66
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR