9-Heptadecanone ≥98%
Dodavatel: Thermo Scientific
L01630.06
L01630.14
L01630.06EA
2230
CZK
L01630.06
L01630.14
9-Heptadecanone ≥98%
9-Heptadecanone
Purity: 98%
CAS No.: 540-08-9
Molecular Formula: C17H34O
Formula Weight: 254.46
Melting Point: 51-53°
Boiling Point: 250-253°
MDL No.: MFCD00009573
Vzorec:
C₁₇H₃₄O MW: 254,46 g/mol Bod varu: 250…253 °C Bod tání: 51…53 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009573 CAS číslo: 540-08-9 EINECS: 208-734-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...