3-Nonanone ≥98%
Dodavatel: Thermo Scientific
Varování
|
A19878.22
A19878.14
A19878.22EA
5190
CZK
A19878.22
A19878.14
3-Nonanone ≥98%
3-Nonanone
Purity: 98%
CAS No.: 925-78-0
Molecular Formula: C9H18O
Formula Weight: 142.24
Flash Point: 63°(145°F)
Refractive Index: 1.4200
Melting Point: -8°
Boiling Point: 187-188°
Beilstein Reference: 1700447
MDL No.: MFCD00009541
Vzorec:
C₉H₁₈O MW: 142,24 g/mol Bod varu: 187…188 °C Bod tání: –8 °C Hustota: 0,825 Bod vzplanutí: 63 °C (145 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009541 CAS číslo: 925-78-0 EINECS: 213-125-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...