Tisk…

2-Nonanone ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: Methyl heptyl ketone, Heptyl methyl ketone

A11781.0B A11781.18 A11781.22
A11781.0BEA 6630 CZK
A11781.0B A11781.18 A11781.22
2-Nonanone ≥98%
2-Nonanone

Purity: 98+% CAS No.: 821-55-6 Molecular Formula: C9H18O Formula Weight: 142.24 Flash Point: 64°(147°F) Refractive Index: 1.4210 Melting Point: -15° Boiling Point: 195° Beilstein Reference: 1743645 MDL No.: MFCD00009553

Vzorec: C₉H₁₈O
MW: 142,24 g/mol
Bod varu: 195 °C
Bod tání: –15 °C
Hustota: 0,826
Bod vzplanutí: 64 °C (147 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009553
CAS číslo: 821-55-6
EINECS: 212-480-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR