Palladium(II) trifluoroacetate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
299685-1G
299685-5G
SIAL299685-1GEA
3910
CZK
SIAL299685-1G
SIAL299685-5G
Palladium(II) trifluoroacetate, Sigma-Aldrich®
Palladium(II) trifluoroacetate
Vzorec:
C₄F₆O₄Pd |
MDL Number:
MFCD00013204 CAS číslo: 42196-31-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...