Palladium(II) trifluoroacetate ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Palladium(II) 2,2,2-trifluoroacetate
39448.03
39448.06
39448.03EA
4190
CZK
39448.03
39448.06
Palladium(II) trifluoroacetate ≥97%
Palladium(II) trifluoroacetate
Purity: 97%
CAS No.: 42196-31-6
Molecular Formula: Pd(O2CCF3)2
Formula Weight: 332.44
Physical Form: Powder
Sensitivity: Hygroscopic
MDL No.: MFCD00013204
Vzorec:
C₄F₆O₄Pd MW: 332.45 Bod tání: 210 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00013204 CAS číslo: 42196-31-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...