Tisk…

Potassium trifluoroacetate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Trifluoroacetic acid potassium salt, Potassium 2,2,2-trifluoroacetate

M-Clarity™ quality level = MQ200

281883-100G 281883-25G
SIAL281883-100GEA 4170 CZK
SIAL281883-100G SIAL281883-25G
Potassium trifluoroacetate, Sigma-Aldrich®
Potassium trifluoroacetate
Vzorec: C₂F₃KO₂
MDL Number: MFCD00013215
CAS číslo: 2923-16-2
EINECS: 220-877-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR