Tisk…

Potassium trifluoroacetate ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Trifluoroacetic acid potassium salt, Potassium 2,2,2-trifluoroacetate

A17398.14 A17398.22
A17398.14EA 1350 CZK
A17398.14 A17398.22
Potassium trifluoroacetate ≥98%
Potassium trifluoroacetate

Purity: 98% CAS No.: 2923-16-2 Molecular Formula: CF3CO2K Formula Weight: 152.12 pH: 7 Melting Point: 142-145° Beilstein Reference: 3717603 UN No.: 2811 MDL No.: MFCD00013215

Vzorec: C₂F₃KO₂
Bod tání: 142…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00013215
CAS číslo: 2923-16-2
EINECS: 220-877-5
UN: 2811
ADR: 6.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR