Thiobenzamide, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Benzothioamide
M-Clarity™ quality level = MQ200
148229-10G
148229-50G
SIAL148229-10GEA
0
CZK
SIAL148229-10G
SIAL148229-50G
Thiobenzamide, Sigma-Aldrich®
Thiobenzamide
Vzorec:
C₇H₇NS MW: 137,21 g/mol Bod varu: 245 °C (1013 hPa) Bod tání: 113…116 °C |
MDL Number:
MFCD00008060 CAS číslo: 2227-79-4 EINECS: 218-765-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...