křemík ≥99.99% (metals basis), crystalline powder -100+200 mesh
Dodavatel: Thermo Fisher Scientific
|
Varování
|
45609.18
45609.30
45609.18EA
4950
CZK
45609.18
45609.30
křemík ≥99.99% (metals basis), crystalline powder -100+200 mesh
křemík
Purity: 99.99%
CAS No.: 7440-21-3
Appearance: White
Odour: Odourless
UN No.: 1346
MDL No.: MFCD00085311
Vzorec:
Si MW: 28.09 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00085311 CAS číslo: 7440-21-3 EINECS: 231-130-8 UN: 1346 ADR: 4.1 III Merck Index: 12,08635 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...