Tisk…

Ethyl-akrylát ≥99% stabilised

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Acrylic acid-ethyl ester

Stabilization: Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)

A15493.AE A15493.AP
A15493.AEEA 1080 CZK
A15493.AE A15493.AP
Ethyl-akrylát ≥99% stabilised
Ethyl-akrylát

Stabilized with ca 20 ppm 4-methoxyphenol


Purity: 99% CAS No.: 140-88-5 Molecular Formula: C5H8O2 Formula Weight: 100.12 Storage Temperature: 2°C to 8°C Flash Point: -1°(30°F) Refractive Index: 1.4060 Odour: Pungent Melting Point: -71° Boiling Point: 99-100° Beilstein Reference: 773866 Merck Reference: 14,3759 UN No.: 1917 MDL No.: MFCD00009188

Vzorec: C₅H₈O₂
MW: 100.12
Bod varu: 99…100 °C
Bod tání: –71 °C
Hustota: 0.924
Bod vzplanutí: –1 °C (30 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00009188
CAS číslo: 140-88-5
EINECS: 205-438-8
UN: 1917
ADR: 3 II
Merck Index: 14,3759

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR