Tisk…

Tributylphenyltin, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Tri-n-butylphenyltin, Tributylphenylstannane

374601-10G 374601-1G
SIAL374601-10GEA 4440 CZK
SIAL374601-10G SIAL374601-1G
Tributylphenyltin, Sigma-Aldrich®
Tributylphenyltin
Vzorec: C₁₈H₃₂Sn
MDL Number: MFCD00134394
CAS číslo: 960-16-7
UN: 2788
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR