Tisk…

Ethyl nitroacetate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

192333-25G 192333-5G
SIAL192333-25GEA 9190 CZK
SIAL192333-25G SIAL192333-5G
Ethyl nitroacetate, Sigma-Aldrich®
Ethyl nitroacetate
Vzorec: C₄H₇NO₄
Bod varu: 93…95 °C (13,3 hPa)
Hustota: 1,226 g/cm³ (0 °C)
MDL Number: MFCD00007403
CAS číslo: 626-35-7
EINECS: 210-944-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR