Tisk…

Oxid neodymitý ≥99.999% (REO, rare earth oxide basis), REacton®

Dodavatel: Thermo Fisher Scientific

Synonyms: Dineodymium trioxide, Neodymium(III)-oxide, Neodymium oxide

45566.14 45566.22
45566.14EA 10850 CZK
45566.14 45566.22
Oxid neodymitý ≥99.999% (REO, rare earth oxide basis), REacton®
Oxid neodymitý

Purity: 99.999% CAS No.: 1313-97-9 Molecular Formula: Nd2O3 Formula Weight: 336.48 Physical Form: Powder Density: 7.24 Appearance: Pale blue Melting Point: 2272° Merck Reference: 14,6450 MDL No.: MFCD00011134

Vzorec: Nd₂O₃
MW: 336.48
Bod varu: 3760
Bod tání: 2272 °C
Hustota: 7.24
Storage Temperature: Ambient
MDL Number: MFCD00011134
CAS číslo: 1313-97-9
EINECS: 215-214-1
Merck Index: 14,6450

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR