Sodium metaborate tetrahydrate, Sigma-Aldrich®
Dodavatel: Merck
S0251-1KG
S0251-500G
SIALS0251-1KGEA
1920
CZK
SIALS0251-1KG
SIALS0251-500G
Sodium metaborate tetrahydrate, Sigma-Aldrich®
Sodium metaborate tetrahydrate
Vzorec:
BNaO₂·4H₂O MW: 137.86 Bod tání: 57 |
MDL Number:
MFCD00149205 CAS číslo: 10555-76-7 EINECS: 231-891-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...