Tisk…

Methoxyacetyl chloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

M9653-10G M9653-1G
SIALM9653-10GEA 2190 CZK
SIALM9653-10G SIALM9653-1G
Methoxyacetyl chloride, Sigma-Aldrich®
Methoxyacetyl chloride
Vzorec: C₃H₅ClO₂
MW: 108.52
MDL Number: MFCD00000728
CAS číslo: 38870-89-2
EINECS: 254-169-2
UN: 2920
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR