Tisk…

Lithium nitrate hydrate ≥99.999% (metals basis), Puratronic®

Dodavatel: Thermo Scientific
Nebezpečí

10742.09 10742.18
10742.09EA 2340 CZK
10742.09 10742.18
Lithium nitrate hydrate ≥99.999% (metals basis), Puratronic®
Lithium nitrate hydrate

Lithium nitrate, anhydrous is used in the manufacture of red-colored fireworks and flares. has been proposed as a medium to store heat collected from the sun for cooking. A Fresnel lens would be used to melt solid lithium nitrate, which would then function as a solar battery, allowing heat to be redistributed later by convection


Purity: 99.999% CAS No.: 7790-69-4 Molecular Formula: LiNO3.xH2O (x~0.5) Formula Weight: 68.95(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic Merck Reference: 14,5536 UN No.: 2722 MDL No.: MFCD00011094

Vzorec: LiNO₃·H₂O
MW: 68,95 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00011094
CAS číslo: 7790-69-4
EINECS: 232-218-9
UN: 2722
ADR: 5.1,III
Merck Index: 14,5536

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR