Ethoxyacetic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Purity: 98%
CAS No.: 627-03-2
Molecular Formula: CH3CH2OCH2CO2H
Formula Weight: 104.11
Flash Point: 97°(206°F)
Refractive Index: 1.4190
Odour: Pungent
Boiling Point: 207°
Beilstein Reference: 1743037
UN No.: 3265
MDL No.: MFCD00004310
Vzorec:
C₄H₈O₃ Bod varu: 207 °C Hustota: 1,102 Bod vzplanutí: 97 °C (206 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004310 CAS číslo: 627-03-2 EINECS: 210-978-2 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...