Tisk…

Tributylvinyltin, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Tri-n-butylvinyltin, Tributylvinylstannane

M-Clarity™ quality level = MQ100

95085-25ML 95085-5ML
SIAL95085-25MLEA 0 CZK
SIAL95085-25ML SIAL95085-5ML
Tributylvinyltin, Sigma-Aldrich®
Tributylvinyltin
Vzorec: C₁₄H₃₀Sn
MW: 317,1 g/mol
Bod varu: 73…74 °C (0,1 mmHg)
Hustota: 1,085 g/cm³ (25 °C)
MDL Number: MFCD00009421
CAS číslo: 7486-35-3
EINECS: 231-291-4
UN: 2788
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR