Tisk…
14-Heptacosanone ≥97%
Dodavatel: Thermo Fisher Scientific
B20191.06 B20191.14
B20191.06EA 2320 CZK
B20191.06 B20191.14
14-Heptacosanone ≥97%
14-Heptacosanone
Purity: 97% CAS No.: 542-50-7 Molecular Formula: C27H54O Formula Weight: 394.73 Melting Point: 76-79° Beilstein Reference: 1796794
Vzorec: C₂₇H₅₄O
MW: 394.73
Bod tání: 76…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00026598
CAS číslo: 542-50-7
EINECS: 208-815-5