Isopropenyl acetate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
117781-1L
117781-250ML
SIAL117781-1LEA
2120
CZK
SIAL117781-1L
SIAL117781-250ML
Isopropenyl acetate, Sigma-Aldrich®
Isopropenyl acetate
Vzorec:
C₅H₈O₂ MW: 100,12 g/mol Bod varu: 94 °C (1013 hPa) Bod tání: –93 °C Hustota: 0,9188 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008709 CAS číslo: 108-22-5 EINECS: 203-562-7 UN: 2403 ADR: 3,II Merck Index: 13,05223 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...