Tisk…

Bromomethyl acetate, Sigma-Aldrich®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ200

303208-10G 303208-1G
SIAL303208-10GEA 13180 CZK
SIAL303208-10G SIAL303208-1G
Bromomethyl acetate, Sigma-Aldrich®
Bromomethyl acetate
Vzorec: H₃CCOOCH₂Br
Storage Temperature: Refrigerator
MDL Number: MFCD00000170
CAS číslo: 590-97-6
EINECS: 209-696-2
UN: 3272
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR