Tisk…

4-Bromobutyl acetate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Acetic acid 4-bromobutyl ester

M-Clarity™ quality level = MQ100

227579-10G 227579-50G
SIAL227579-10GEA 1890 CZK
SIAL227579-10G SIAL227579-50G
4-Bromobutyl acetate, Sigma-Aldrich®
4-Bromobutyl acetate
Vzorec: C₆H₁₁BrO₂
MW: 195,06 g/mol
MDL Number: MFCD00000263
CAS číslo: 4753-59-7
EINECS: 225-277-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR