Palladium(II) acetate (45,9 - 48,4% Pd)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Palladium diacetate, Palladium (II) acetate
10516.03
10516.06
10516.14
10516.03EA
5630
CZK
10516.03
10516.06
10516.14
Palladium(II) acetate (45,9 - 48,4% Pd)
Palladium(II) acetate
CAS No.: 3375-31-3
Molecular Formula: C12H18O12Pd3
Formula Weight: 673.46
Physical Form: Needles
Appearance: Red-brown
Melting Point: ca 205° dec.
MDL No.: MFCD00012453
Carbonylation, oxidation and C-C bond formation
Vzorec:
C₁₂H₁₈O₁₂Pd₃ MW: 224.51 Bod tání: ∼ 205 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00012453 CAS číslo: 3375-31-3 EINECS: 222-164-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...