Tisk…

Manganese(III) acetate dihydrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Acetic acid manganese (III) salt dihydrate, Manganese triacetate dihydrate

M-Clarity™ quality level = MQ100

215880-100G 215880-25G 215880-5G
SIAL215880-100GEA 19710 CZK
SIAL215880-100G SIAL215880-25G SIAL215880-5G
Manganese(III) acetate dihydrate, Sigma-Aldrich®
Manganese(III) acetate dihydrate
Vzorec: (H₃CCOO)₂Mn·2H₂O
MW: 209,06 g/mol
MDL Number: MFCD00150022
CAS číslo: 19513-05-4
EINECS: 213-602-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR