Tisk…

Cerium triacetate hydrate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

367753-250G 367753-50G
SIAL367753-250GEA 7700 CZK
SIAL367753-250G SIAL367753-50G
Cerium triacetate hydrate, Sigma-Aldrich®
Cerium triacetate hydrate
Vzorec: C₆H₉CeO₆·H₂O
MDL Number: MFCD00150533
CAS číslo: 206996-60-3
EINECS: 208-654-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR