Formaldehyde diethyl acetal ≥99% stabilizovaný
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Ethylal, Diethoxymethane
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
B23134.AE
B23134.AP
B23134.AEEA
1600
CZK
B23134.AE
B23134.AP
Formaldehyde diethyl acetal ≥99% stabilizovaný
Formaldehyde diethyl acetal
Stabilized with 100 ppm BHT
Purity: 99%
CAS No.: 462-95-3
Molecular Formula: CH3CH2OCH2OCH2CH3
Formula Weight: 104.15
Flash Point: -5°(23°F)
Physical Form: Liquid
Density: 0.831
Refractive Index: 1.3740
Melting Point: -66°
Boiling Point: 87-88°
Beilstein Reference: 1697253
UN No.: 2373
MDL No.: MFCD00009246
Vzorec:
C₅H₁₂O₂ MW: 104,15 g/mol Bod varu: 87…88 °C Bod tání: –66 °C Hustota: 0,831 Bod vzplanutí: –5 °C (23 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009246 CAS číslo: 462-95-3 EINECS: 207-330-6 UN: 2373 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...