Tisk…

Cytochalasin B (from Dreschslera dematioidea), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 7(S),20(R)-Dihydroxy-16(R)-methyl-10-phenyl-24-oxa[14]cytochalasa-6(12),13(E),21(E)-triene-1,23-dione

M-Clarity™ quality level = MQ300

C6762-1MG C6762-25MG C6762-50MG C6762-5MG C6762-10MG
SIALC6762-1MGEA 4180 CZK
SIALC6762-1MG SIALC6762-25MG SIALC6762-50MG SIALC6762-5MG SIALC6762-10MG
Cytochalasin B (from Dreschslera dematioidea), Sigma-Aldrich®
Cytochalasin B
Vzorec: C₂₉H₃₇NO₅
MW: 479,62 g/mol
MDL Number: MFCD00077704
CAS číslo: 14930-96-2
EINECS: 239-000-2
UN: 1544
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR