Tisk…

Cyclohexyltriphenylphosphonium bromide ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A14298.06 A14298.14 A14298.22
A14298.06EA 1070 CZK
A14298.06 A14298.14 A14298.22
Cyclohexyltriphenylphosphonium bromide ≥98%
Cyclohexyltriphenylphosphonium bromide

Purity: 98+% CAS No.: 7333-51-9 Molecular Formula: C24H26BrP Formula Weight: 425.36 Melting Point: 265-272° Beilstein Reference: 3581012

Vzorec: C₂₄H₂₆BrP
Bod tání: 265…272 °C
Storage Temperature: Ambient
MDL Number: MFCD00031667
CAS číslo: 7333-51-9
EINECS: 230-834-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR