Cyclohexylacetic acid ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Cyclohexaneacetic acid
H31986.06
H31986.14
H31986.06EA
1220
CZK
H31986.06
H31986.14
Cyclohexylacetic acid ≥98%
Cyclohexylacetic acid
Purity: 98%
CAS No.: 5292-21-7
Molecular Formula: C8H14O2
Formula Weight: 142.2
Melting Point: 28-32°
Boiling Point: 242-244°
MDL No.: MFCD00001518
Vzorec:
C₈H₁₄O₂ MW: 142,2 g/mol Bod varu: 242…244 °C Bod tání: 28…32 °C Hustota: 1,007 Storage Temperature: Ambient |
MDL Number:
MFCD00001518 CAS číslo: 5292-21-7 EINECS: 226-132-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...