Tisk…

Benzyl cyanoacetate ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L06644.14 L06644.22 AB143945.0100
L06644.14EA 2930 CZK
L06644.14 L06644.22 ABCRAB143945.0100
Benzyl cyanoacetate ≥97%
Benzyl cyanoacetate

Purity: 97% CAS No.: 14447-18-8 Molecular Formula: NCCH2CO2CH2C6H5 Formula Weight: 175.18 Flash Point: 100°(212°F) Refractive Index: 1.5195 Odour: Odourless Melting Point: 26-30° Boiling Point: 140°/0.8mm Beilstein Reference: 2096578 MDL No.: MFCD00051719

Vzorec: C₁₀H₉NO₂
Bod varu: 140 °C (0,8 mmHg)
Bod tání: 26…30 °C
Bod vzplanutí: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00051719
CAS číslo: 14447-18-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR