Coronene, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Hexabenzobenzene
MQ100
C84801-100MG
C84801-500MG
SIALC84801-100MGEA
1920
CZK
SIALC84801-100MG
SIALC84801-500MG
Coronene, Sigma-Aldrich®
Coronene
Vzorec:
C₂₄H₁₂ MW: 300.36 Bod varu: 525 Bod tání: 437…440 °C Hustota: 1.371 g/cm³ (25 °C) |
MDL Number:
MFCD00004134 CAS číslo: 191-07-1 EINECS: 205-881-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...