Tisk…

Diaminomaleonitrile 98%

Dodavatel: Thermo Scientific
Nebezpečí

B24328.14 B24328.22
B24328.14EA 1220 CZK
B24328.14 B24328.22
Diaminomaleonitrile 98%
Diaminomaleonitrile

Purity: 98% CAS No.: 1187-42-4 Molecular Formula: C4H4N4 Formula Weight: 108.10 Melting Point: 178-182° dec. Beilstein Reference: 878333 UN No.: 3439 MDL No.: MFCD00001870

Vzorec: C₄H₄N₄
MW: 108,1 g/mol
Bod varu: 445 °C (1013 hPa)
Bod tání: 178…182 °C (rozkl.)
Hustota: 1,360
Storage Temperature: Ambient
MDL Number: MFCD00001870
CAS číslo: 1187-42-4
EINECS: 214-697-6
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR