Methyl benzenesulphonate ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
B24515.14
B24515.22
B24515.36
B24515.14EA
1160
CZK
B24515.14
B24515.22
B24515.36
Methyl benzenesulphonate ≥98%
Methyl benzenesulphonate
Purity: 98%
CAS No.: 80-18-2
Molecular Formula: C7H8O3S
Formula Weight: 172.20
Flash Point: 143°(289°F)
Refractive Index: 1.5165
Melting Point: -4°
Boiling Point: 280°
Beilstein Reference: 908448
UN No.: 3265
MDL No.: MFCD00014737
Vzorec:
C₇H₈O₃S MW: 172,2 g/mol Bod varu: 280 °C Bod tání: –4 °C Hustota: 1,269 Bod vzplanutí: 143 °C (289 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014737 CAS číslo: 80-18-2 EINECS: 201-256-8 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...