Tisk…

Benzamidine hydrochloride, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Benzimidamide hydrochloride, Benzamidinium chloride, Phenylamidine hydrochloride, Benzene-1-carboximidamide hydrochloride

M-Clarity™ quality level = MQ200

434760-25G 434760-5G
SIAL434760-25GEA 19830 CZK
SIAL434760-25G SIAL434760-5G
Benzamidine hydrochloride, Sigma-Aldrich®
Benzamidine hydrochloride
Vzorec: C₇H₉ClN₂
Bod tání: 174…179 °C
MDL Number: MFCD00066285
CAS číslo: 1670-14-0
EINECS: 216-795-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR