Tisk…

Benzamidine hydrochloride 99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Benzimidamide hydrochloride, Benzamidinium chloride, Phenylamidine hydrochloride, Benzene-1-carboximidamide hydrochloride

401790250 401791000 401790050 401795000
ACRO401790250EA 3580 CZK
ACRO401790250 ACRO401791000 ACRO401790050 ACRO401795000
Benzamidine hydrochloride 99%
Benzamidine hydrochloride
Vzorec: C₇H₉ClN₂
Bod tání: 167…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00066285
CAS číslo: 1670-14-0
EINECS: 216-795-4

Objednat

Výsledky specifikačního testu

Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
Water ≤1.0 % (K.F.)
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR