Tisk…

Dexamethasone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Dexamethasonum, 9-Fluor-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadien-3,20-dion

M-Clarity™ quality level = MQ200

D1756-100MG D1756-25MG D1756-1G D1756-500MG D1756-5G
SIALD1756-100MGEA 4660 CZK
SIALD1756-100MG SIALD1756-25MG SIALD1756-1G SIALD1756-500MG SIALD1756-5G
Dexamethasone, Sigma-Aldrich®
Dexamethasone
Vzorec: C₂₂H₂₉FO₅
Bod tání: 268…271 °C
MDL Number: MFCD00064136
CAS číslo: 50-02-2
EINECS: 200-003-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR